Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) 95%

Supplier: Ambeed
Warning

A758916-1G A758916-250MG
77599-012EA 86.1 USD
77599-012 77599-014
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) 95%
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate)

Precaución: for Research Use Only

Formula: C₃₀H₂₄CoF₁₂N₆P₂
MW: 817.43 g/mol
CAS Number: 79151-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR