N-(5-(6-Fluoro-8-((4-(2-hydroxypropan-2-yl)piperidin-1-yl)methyl)-2-morpholinoquinazolin-4-yl)-2-methoxypyridin-3-yl)methanesulfonamide 99%

Supplier: Ambeed
Warning

A724078-1MG A724078-10MG A724078-5MG
77598-812EA 29.73 USD
77598-812 77622-582 77622-584
N-(5-(6-Fluoro-8-((4-(2-hydroxypropan-2-yl)piperidin-1-yl)methyl)-2-morpholinoquinazolin-4-yl)-2-methoxypyridin-3-yl)methanesulfonamide 99%
N-(5-(6-Fluoro-8-((4-(2-hydroxypropan-2-yl)piperidin-1-yl)methyl)-2-morpholinoquinazolin-4-yl)-2-methoxypyridin-3-yl)methanesulphonamide

Precaución: for Research Use Only

Formula: C₂₈H₃₇FN₆O₅S
MW: 588.6940 g/mol
MDL Number: MFCD31544346
CAS Number: 1702816-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR