(1S,2S)-2-Iodocyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

A691008-100MG A691008-1G A691008-250MG
77598-706EA 162.98 USD
77598-706 77598-708 77598-710
(1S,2S)-2-Iodocyclopropanecarboxylic acid 97%
(1S,2S)-2-Iodocyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅IO₂
MW: 211.9858 g/mol
MDL Number: MFCD28016082
CAS Number: 1932315-23-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR