α-Glycerolphosphate 35%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dihydroxypropyl dihydrogen phosphate, 2,3-Hydroxy-1-propyl dihydrogen phosphate, Glycerol 3-phosphate

A650378-5G A650378-25G
77598-656EA 17.43 USD
77598-656 77627-300
α-Glycerolphosphate 35%
α-Glycerolphosphate

Precaución: for Research Use Only

Formula: C₃H₉O₆P
MW: 172.07 g/mol
MDL Number: MFCD00152510
CAS Number: 57-03-4
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR