(T-4)-Copper(1+), bis[2,9-bis(4-methoxyphenyl)-1,10-phenanthroline-κN1,κN10]-, chloride 95%

Supplier: Ambeed
Warning

A489217-100MG A489217-1G A489217-250MG
77598-514EA 98.4 USD
77598-514 77598-516 77598-518
(T-4)-Copper(1+), bis[2,9-bis(4-methoxyphenyl)-1,10-phenanthroline-κN1,κN10]-, chloride 95%
(T-4)-Copper(1+), bis[2,9-bis(4-methoxyphenyl)-1,10-phenanthroline-κN1,κN10]-, chloride

Precaución: for Research Use Only

Formula: C₅₂H₄₀ClCuN₄O₄
CAS Number: 1392109-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR