Ir[dF(CF3)ppy]2[(CH2CH2CO2H-Me)ppy]PF6 95%

Supplier: Ambeed
Warning

A2085689-100MG A2085689-250MG A2085689-50MG
77598-310EA 292.13 USD
77598-310 77598-312 77598-314
Ir[dF(CF3)ppy]2[(CH2CH2CO2H-Me)ppy]PF6 95%
Ir[dF(CF3)ppy]2[(CH2CH2CO2H-Me)ppy]PF6

Precaución: for Research Use Only

Formula: C₃₈H₂₄F₁₆IrN₄O₂P
CAS Number: 2725035-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR