6-(2-(Diethylamino)ethoxy)-N,N-dimethylbenzo[d]thiazol-2-amine hydrochloride 98%

Supplier: Ambeed
Warning

A2021658-1MG A2021658-10MG A2021658-5MG
77598-298EA 76.88 USD
77598-298 77626-028 77626-030
6-(2-(Diethylamino)ethoxy)-N,N-dimethylbenzo[d]thiazol-2-amine hydrochloride 98%
6-(2-(Diethylamino)ethoxy)-N,N-dimethylbenzo[d]thiazol-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄ClN₃OS
MW: 329.8886 g/mol
CAS Number: 17140-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR