RMC-6236 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-N-[(2R,14S,18S)-1-Ethyl-18,19,20,21-tetrahydro-2-[2-[(1S)-1-methoxyethyl]-5-(4-methyl-1-piperazinyl)-3-pyridinyl]-25,25-dimethyl-15,22-dioxo-17H-5,3-([4,2]-endo-thiazolopropano[1,3]-endo-pyridazinomethanoxypropano)-1H-indol-14-yl]-2-methyl-cyclopropanecarboxamide

A1727701-1MG A1727701-10MG A1727701-5MG
77598-222EA 904.05 USD
77598-222 77625-804 77625-806
RMC-6236 98%
RMC-6236

Precaución: for Research Use Only

Formula: C₄₄H₅₈N₈O₅S
MW: 811.05 g/mol
CAS Number: 2765081-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR