4-Ethynyl-2,2'-bipyridine 98%

Supplier: Ambeed
Warning

A1660702-100MG A1660702-250MG
77598-128EA 128.13 USD
77598-128 77598-130
4-Ethynyl-2,2'-bipyridine 98%
4-Ethynyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₂H₈N₂
MW: 180.2053 g/mol
MDL Number: MFCD32709717
CAS Number: 146548-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR