ASN03576800 98%

Supplier: Ambeed
Warning

Synonyms: 2-((2-(Benzo[d][1,3]dioxol-5-ylamino)-2-oxoethyl)sulphinyl)acetic acid

A1615363-1MG A1615363-10MG A1615363-5MG
77598-116EA 30.75 USD
77598-116 77625-564 77625-566
ASN03576800 98%
ASN03576800

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₆S
MW: 285.2731 g/mol
MDL Number: MFCD03274882
CAS Number: 957513-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR