4-Methyl-3-((6-(methylamino)pyrimidin-4-yl)oxy)-N-(3-(4-methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl)benzamide 98%
Supplier: Ambeed
|
Warning
|
A1595804-1MG
A1595804-5MG
77598-080EA
100.45
USD
77598-080
77625-482
4-Methyl-3-((6-(methylamino)pyrimidin-4-yl)oxy)-N-(3-(4-methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl)benzamide 98%
4-Methyl-3-((6-(methylamino)pyrimidin-4-yl)oxy)-N-(3-(4-methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl)benzamide
Precaución: for Research Use Only
Formula:
C₂₅H₂₇F₃N₆O₂ MW: 500.5161 g/mol |
MDL Number:
MFCD31689241 CAS Number: 1620820-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...