(S)-4-Methyl-5-((4-((2-(methylamino)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1-(2-(4-(methylsulfonyl)piperazin-1-yl)propyl)-1H-indole-2-carbonitrile 97%

Supplier: Ambeed
Warning

A1550156-1MG A1550156-10MG
77598-030EA 235.75 USD
77598-030 77625-394
(S)-4-Methyl-5-((4-((2-(methylamino)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1-(2-(4-(methylsulfonyl)piperazin-1-yl)propyl)-1H-indole-2-carbonitrile 97%
(S)-4-Methyl-5-((4-((2-(methylamino)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1-(2-(4-(methylsulphonyl)piperazin-1-yl)propyl)-1H-indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₃₃H₄₂F₃N₉O₂S₂
MW: 717.8700 g/mol
CAS Number: 2134675-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR