(2S)-Isopropyl 2-(((((2R,3R,4R,5R)-5-(2-amino-6-(methylamino)-9H-purin-9-yl)-4-fluoro-3-hydroxy-4-methyltetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate hemisulfate 98%

Supplier: Ambeed
Danger

A1501228-1MG A1501228-100MG A1501228-25MG A1501228-50MG
77597-770EA 85.08 USD
77597-770 77875-735 77875-736 77875-737
(2S)-Isopropyl 2-(((((2R,3R,4R,5R)-5-(2-amino-6-(methylamino)-9H-purin-9-yl)-4-fluoro-3-hydroxy-4-methyltetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate hemisulfate 98%
(2S)-Isopropyl 2-(((((2R,3R,4R,5R)-5-(2-amino-6-(methylamino)-9H-purin-9-yl)-4-fluoro-3-hydroxy-4-methyltetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate hemisulfate

Precaución: for Research Use Only

Formula: C₄₈H₆₈F₂N₁₄O₁₈P₂S
MW: 1261.1459 g/mol
CAS Number: 2241337-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR