[N-[Phenyl[2-[[[(1S,2R)-1-(phenylmethyl)-2-pyrrolidinyl-κN]carbonyl]amino-κN]phenyl]methylene]glycinato(2-)-κN,κO]nickel 98%

Supplier: Ambeed
Danger

A1472922-1G A1472922-250MG A1472922-5G
77597-418EA 58.43 USD
77597-418 77597-420 77597-422
[N-[Phenyl[2-[[[(1S,2R)-1-(phenylmethyl)-2-pyrrolidinyl-κN]carbonyl]amino-κN]phenyl]methylene]glycinato(2-)-κN,κO]nickel 98%
[N-[Phenyl[2-[[[(1S,2R)-1-(phenylmethyl)-2-pyrrolidinyl-κN]carbonyl]amino-κN]phenyl]methylene]glycinato(2-)-κN,κO]nickel

Precaución: for Research Use Only

Formula: C₂₇H₂₅N₃NiO₃
MW: 498.1991 g/mol
MDL Number: MFCD18910208
CAS Number: 172924-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR