N,N-Dimethyl-2-((2-oxo-4-(o-tolyl)-2H-chromen-7-yl)oxy)propanamide 98%

Supplier: Ambeed
Warning

A1466976-1MG A1466976-100MG A1466976-250MG A1466976-25MG A1466976-50MG
77597-378EA 56.38 USD
77597-378 77875-038 77875-039 77875-040 77875-041
N,N-Dimethyl-2-((2-oxo-4-(o-tolyl)-2H-chromen-7-yl)oxy)propanamide 98%
N,N-Dimethyl-2-((2-oxo-4-(o-tolyl)-2H-chromen-7-yl)oxy)propanamide

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
CAS Number: 2304621-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR