Chloro[(1,3,5,7-tetramethyl-5-phenyl-2,4,8-trioxa-6-phosphaadamantane)-2-(2-aminobiphenyl)]palladium(II) 98%

Supplier: Ambeed
Warning

A1466043-100MG A1466043-250MG
77597-368EA 86.1 USD
77597-368 77597-370
Chloro[(1,3,5,7-tetramethyl-5-phenyl-2,4,8-trioxa-6-phosphaadamantane)-2-(2-aminobiphenyl)]palladium(II) 98%
Chloro[(1,3,5,7-tetramethyl-5-phenyl-2,4,8-trioxa-6-phosphaadamantane)-2-(2-aminobiphenyl)]palladium(II)

Precaución: for Research Use Only

Formula: C₂₈H₃₁ClNO₃PPd
MW: 602.3974 g/mol
MDL Number: MFCD22572665
CAS Number: 1350851-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR