Zinc 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine 98%

Supplier: Ambeed
Warning

A1465809-100MG A1465809-250MG A1465809-1G
77597-364EA 110.7 USD
77597-364 77597-366 77875-031
Zinc 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine 98%
Zinc 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine

Precaución: for Research Use Only

Formula: C₃₂H₆F₁₆N₈Zn
MW: 871.8200 g/mol
MDL Number: MFCD00216667
CAS Number: 31396-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR