2,2-Bis((3aR,8aS)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A1452001-100MG A1452001-250MG
77597-220EA 144.53 USD
77597-220 77597-222
2,2-Bis((3aR,8aS)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)acetonitrile 97%
2,2-Bis((3aR,8aS)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃O₂
MW: 355.3893 g/mol
CAS Number: 1251471-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR