(S)-5,5'-Bis(dicyclohexylphosphino)-4,4'-bibenzo[d][1,3]dioxole 97%

Supplier: Ambeed
Warning

A1437935-100MG A1437935-250MG A1437935-1G
77597-180EA 142.48 USD
77597-180 77597-182 77874-797
(S)-5,5'-Bis(dicyclohexylphosphino)-4,4'-bibenzo[d][1,3]dioxole 97%
(S)-5,5'-Bis(dicyclohexylphosphino)-4,4'-bibenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₃₈H₅₂O₄P₂
CAS Number: 958300-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR