(4-Amino-2-(isopropylamino)thiazol-5-yl)(phenyl)methanone 98%

Supplier: Ambeed
Warning

A1398865-1MG A1398865-2MG A1398865-5MG
77597-138EA 29.73 USD
77597-138 77622-028 77622-030
(4-Amino-2-(isopropylamino)thiazol-5-yl)(phenyl)methanone 98%
(4-Amino-2-(isopropylamino)thiazol-5-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅N₃OS
MW: 261.3427 g/mol
MDL Number: MFCD12034481
CAS Number: 204188-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR