(S)-1,4-Oxazepan-6-ol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A1368765-1G
A1368765-100MG
A1368765-250MG
77596-992EA
481.75
USD
77596-992
77621-854
77621-856
(S)-1,4-Oxazepan-6-ol hydrochloride 98%
(S)-1,4-Oxazepan-6-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂ |
MDL Number:
MFCD31801282 CAS Number: 2708343-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...