tert-Butyl ((S)-3-methyl-1-(((S)-1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxopropan-2-yl)amino)-1-oxobutan-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A1365410-100MG A1365410-1G A1365410-250MG
77596-956EA 211.15 USD
77596-956 77596-958 77621-828
tert-Butyl ((S)-3-methyl-1-(((S)-1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxopropan-2-yl)amino)-1-oxobutan-2-yl)carbamate 98%
tert-Butyl ((S)-3-methyl-1-(((S)-1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxopropan-2-yl)amino)-1-oxobutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₄O₉
MW: 558.5803 g/mol
MDL Number: MFCD28557281
CAS Number: 1884578-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR