4-(2-Amino-4-methyl-3-(2-methylquinolin-6-yl)benzoyl)-1-methyl-2,5-diphenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A1365024-1MG
A1365024-5MG
77596-942EA
38.95
USD
77596-942
77621-812
4-(2-Amino-4-methyl-3-(2-methylquinolin-6-yl)benzoyl)-1-methyl-2,5-diphenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride 98%
4-(2-Amino-4-methyl-3-(2-methylquinolin-6-yl)benzoyl)-1-methyl-2,5-diphenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride
Precaución: for Research Use Only
Formula:
C₃₄H₂₉ClN₄O₂ MW: 561.0727 g/mol |
MDL Number:
MFCD30182311 CAS Number: 1883548-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...