3,5,7-Trihydroxy-8-(3-hydroxy-3-methylbutyl)-2-(4-methoxyphenyl)-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

A1364774-1MG A1364774-10MG A1364774-5MG
77596-926EA 19.48 USD
77596-926 77621-806 77621-808
3,5,7-Trihydroxy-8-(3-hydroxy-3-methylbutyl)-2-(4-methoxyphenyl)-4H-chromen-4-one 98%
3,5,7-Trihydroxy-8-(3-hydroxy-3-methylbutyl)-2-(4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₇
MW: 386.3952 g/mol
MDL Number: MFCD25973549
CAS Number: 521-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR