Silver(I) tetrakis(perfluorophenyl)borate 98%

Supplier: Ambeed
Danger

A1353128-100MG A1353128-250MG A1353128-1G
77596-840EA 151.7 USD
77596-840 77596-842 77867-467
Silver(I) tetrakis(perfluorophenyl)borate 98%
Silver(I) tetrakis(perfluorophenyl)borate

Precaución: for Research Use Only

Formula: C₂₄AgBF₂₀
CAS Number: 143607-32-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR