(4R,4'R)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%

Supplier: Ambeed
Warning

A1351971-100MG A1351971-250MG A1351971-1G
77596-824EA 98.4 USD
77596-824 77596-826 77867-447
(4R,4'R)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%
(4R,4'R)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂O₂
MW: 304.4271 g/mol
CAS Number: 2378751-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR