2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26-octaoxa-5-azaoctacosan-28-oic acid 98%

Supplier: Ambeed
Warning

A1329379-1G A1329379-100MG A1329379-250MG
77599-908EA 405.9 USD
77599-908 77596-786 77596-788
2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26-octaoxa-5-azaoctacosan-28-oic acid 98%
2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26-octaoxa-5-azaoctacosan-28-oic acid

Precaución: for Research Use Only

Formula: C₂₁H₄₁NO₁₁
MW: 483.5504 g/mol
MDL Number: MFCD08274604
CAS Number: 141282-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR