(R)-2-((2-Amino-7-fluoropyrido[3,2-d]pyrimidin-4-yl)amino)-2-methylhexan-1-ol 98%

Supplier: Ambeed
Warning

A1250870-1MG A1250870-10MG A1250870-5MG
77596-558EA 141.45 USD
77596-558 77621-246 77621-248
(R)-2-((2-Amino-7-fluoropyrido[3,2-d]pyrimidin-4-yl)amino)-2-methylhexan-1-ol 98%
(R)-2-((2-Amino-7-fluoropyrido[3,2-d]pyrimidin-4-yl)amino)-2-methylhexan-1-ol

Precaución: for Research Use Only

Formula: C₁₄H₂₀FN₅O
MW: 293.3399 g/mol
CAS Number: 2004677-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR