9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A1207354-100MG A1207354-250MG
77596-248EA 124.03 USD
77596-248 77596-250
9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate 97%
9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₂H₃₂BF₄NO₄
MW: 581.4054 g/mol
MDL Number: MFCD31381912
CAS Number: 1965330-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR