Manganese(II) stearate 98%

Supplier: Ambeed
Warning

A1166738-25G A1166738-100G
77595-988EA 9.23 USD
77595-988 77866-207
Manganese(II) stearate 98%
Manganese(II) stearate

Precaución: for Research Use Only

Formula: C₃₆H₇₀MnO₄
MW: 621.8767 g/mol
CAS Number: 3353-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR