Di-μ-chlorodichlorobis[(1,2,3,4,5-η)-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl]dicobalt 97%
Supplier: Ambeed
|
Warning
|
A1159503-100MG
A1159503-250MG
77595-954EA
118.9
USD
77595-954
77595-956
Di-μ-chlorodichlorobis[(1,2,3,4,5-η)-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl]dicobalt 97%
Di-μ-chlorodichlorobis[(1,2,3,4,5-η)-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl]dicobalt
Precaución: for Research Use Only
Formula:
C₂₀H₃₀Cl₄Co₂ MW: 530.1300 g/mol |
CAS Number:
82595-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...