Ruthenium-tetrakis(acetonitrile)[2-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl-κN3]phenyl-κC]-hexafluorophosphate 98%

Supplier: Ambeed
Warning

A1157670-100MG A1157670-250MG
77595-936EA 111.73 USD
77595-936 77595-938
Ruthenium-tetrakis(acetonitrile)[2-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl-κN3]phenyl-κC]-hexafluorophosphate 98%
Ruthenium-tetrakis(acetonitrile)[2-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl-κN3]phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₂₃H₂₄F₆N₅OPRu
MW: 632.5038 g/mol
MDL Number: MFCD34646238
CAS Number: 1421679-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR