(Methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC](tricyclohexylphosphine)palladium 98%
Supplier: Ambeed
|
Warning
|
A1155292-100MG
A1155292-250MG
A1155292-1G
A1155292-5G
77595-916EA
64.58
USD
77595-916
77595-918
77620-214
77866-057
(Methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC](tricyclohexylphosphine)palladium 98%
(Methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC](tricyclohexylphosphine)palladium
Precaución: for Research Use Only
Formula:
C₃₂H₄₈NO₃PPdS |
MDL Number:
MFCD30480498 CAS Number: 2195390-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...