(2,2'-Bipyridine)(4-ethynyl-2,2'-bipyridine)Ruthenium hexafluorophosphate 98%

Supplier: Ambeed
Warning

A1150985-100MG A1150985-50MG A1150985-250MG
77595-882EA 261.38 USD
77595-882 77595-884 77865-976
(2,2'-Bipyridine)(4-ethynyl-2,2'-bipyridine)Ruthenium hexafluorophosphate 98%
(2,2'-Bipyridine)(4-ethynyl-2,2'-bipyridine)Ruthenium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₂H₂₄F₁₂N₆P₂Ru
CAS Number: 164357-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR