Iodotris(triphenylphosphine)copper 97%

Supplier: Ambeed
Warning

A1143018-1G A1143018-250MG A1143018-5G
77595-778EA 43.05 USD
77595-778 77619-934 77865-835
Iodotris(triphenylphosphine)copper 97%
Iodotris(triphenylphosphine)copper

Precaución: for Research Use Only

Formula: C₅₄H₄₅CuIP₃
MW: 977.3069 g/mol
CAS Number: 15709-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR