Acridine-9-carbonyl chloride 95%

Supplier: Ambeed
Danger

A982985-1G A982985-250MG A982985-5G
77595-570EA 157.85 USD
77595-570 77595-572 77684-911
Acridine-9-carbonyl chloride 95%
Acridine-9-carbonyl chloride

Precaución: For research use only.

Formula: C₁₄H₈ClNO
MW: 241.6724 g/mol
MDL Number: MFCD08063872
CAS Number: 66074-67-7
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR