(1R,2R,5S)-3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A424184-1G A424184-250MG
77594-308EA 436.65 USD
77594-308 77594-310
(1R,2R,5S)-3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-2-carboxylic acid 97%
(1R,2R,5S)-3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD22536388
CAS Number: 1389359-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR