(±)-Mercaptosuccinic acid 98%

Supplier: Ambeed
Danger

A385415-100G A385415-25G A385415-500G
77594-168EA 71.75 USD
77594-168 77594-170 77601-698
(±)-Mercaptosuccinic acid 98%
(±)-Mercaptosuccinic acid

Precaución: for Research Use Only

Formula: C₄H₆O₄S
MDL Number: MFCD00004860
CAS Number: 70-49-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR