3-Bromo-4-ethylpyridine 97%

Supplier: Ambeed
Warning

A327545-100MG A327545-1G A327545-250MG A327545-5G
77593-934EA 44.08 USD
77593-934 77593-936 77593-938 77872-472
3-Bromo-4-ethylpyridine 97%
3-Bromo-4-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MDL Number: MFCD11036137
CAS Number: 38749-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR