tert-Butyl (3-formylbicyclo[1.1.1]pentan-1-yl)carbamate 97%

Supplier: Ambeed
Warning

A279851-100MG A279851-1G A279851-250MG
77593-722EA 128.13 USD
77593-722 77593-724 77593-726
tert-Butyl (3-formylbicyclo[1.1.1]pentan-1-yl)carbamate 97%
tert-Butyl (3-formylbicyclo[1.1.1]pentan-1-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.258 g/mol
MDL Number: MFCD27987008
CAS Number: 1638771-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR