(2R,2'R)-2,2'-Bipyrrolidine (2R,3R)-2,3-dihydroxysuccinate 97%

Supplier: Ambeed
Warning

A275836-250MG A275836-100MG A275836-1G
77593-702EA 76.88 USD
77593-702 77616-156 77748-529
(2R,2'R)-2,2'-Bipyrrolidine (2R,3R)-2,3-dihydroxysuccinate 97%
(2R,2'R)-2,2'-Bipyrrolidine (2R,3R)-2,3-dihydroxysuccinate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₆
MW: 290.3129 g/mol
MDL Number: MFCD11656040
CAS Number: 137037-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR