Malonaldehydebis(phenylimine) monohydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N1,N3-Diphenylpropane-1,3-diimine hydrochloride

A217866-5G A217866-100G A217866-10G A217866-25G
77593-416EA 23.58 USD
77593-416 77615-828 77615-830 77615-832
Malonaldehydebis(phenylimine) monohydrochloride 98%
Malonaldehydebis(phenylimine) monohydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂·HCl
MW: 258.75 g/mol
MDL Number: MFCD00054634
CAS Number: 123071-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR