2-Amino-6-nitrobenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Nitrobenzothiazol-2-ylamine, 6-Nitro-1,3-benzothiazol-2-amine, 6-Nitrobenzo[d]thiazol-2-amine

A170710-10G A170710-25G A170710-5G
77593-056EA 46.13 USD
77593-056 77593-058 77593-060
2-Amino-6-nitrobenzothiazole 98%
2-Amino-6-nitrobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂S
MW: 195.2 g/mol
MDL Number: MFCD00005786
CAS Number: 6285-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR