Methyl 2-(4-bromo-2,5-difluorophenyl)acetate 98%

Supplier: Ambeed
Warning

A1375186-1G A1375186-5G A1375186-250MG
77592-616EA 80.98 USD
77592-616 77597-052 77621-942
Methyl 2-(4-bromo-2,5-difluorophenyl)acetate 98%
Methyl 2-(4-bromo-2,5-difluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MW: 265.0515 g/mol
MDL Number: MFCD28738330
CAS Number: 1805105-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR