α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: alpha,alpha,alpha',alpha'-Tetramethyl-1,3-benzenedipropionic acid, 3,3'-(1,3-Phenylene)bis(2,2-dimethylpropanoic acid)

A1154290-1G A1154290-250MG A1154290-5G
77592-248EA 46.13 USD
77592-248 77595-902 77620-170
α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid 98%
α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₄
MW: 278.35 g/mol
MDL Number: MFCD08276874
CAS Number: 819050-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR