Emergency Deluge Showers, Speakman®
Supplier: Speakman
SE-237 SE-238
14224-244EA 2885.27 USD
14224-244 14224-246
Emergency Deluge Showers, Speakman®
First Aid Supplies Showers
Emergency deluge showers feature a fully recessed wall activator.
The standard shower has a 25.4 cm (10") chrome-plated flanged showerhead that fits snugly against the ceiling.
The extended shower, designed to accommodate ceilings taller than 2.4 cm (8'), has a chrome-plated showerhead that can be positioned at the showerhead height of 2.4 cm (8').
Información para pedidos: Complete with showerhead, valve and handle, and stainless steel recessed lipped panel. Installation of recessed panel requires a minimum wall thickness of 15.2 cm (6"). Internal wall piping is not supplied and must be provided by customer or plumber.