Phe-Pro

Supplier: Ambeed

A263284-100MG A263284-1G A263284-250MG
77601-128EA 42.03 USD
77601-128 77601-130 77601-132
Phe-Pro
Proteins and Peptides
New Product

Order Now

SPECIFICATIONS

Protein/Peptide Name Phe-Pro
Environmentally Preferable
Protein Synonyms (S)-1-((S)-2-Amino-3-phenylpropanoyl)pyrrolidine-2-carboxylic acid
Species
Source
CAS No. 7669-65-0
MDL No. MFCD00020833
Purity 98%
Molecular Weight 262.3043
Molecular Formula C14H18N2O3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR