Xylene Resistant Resin Ribbons
Supplier: GA INTERNATIONAL INC
807656004280 807656005201 807656004297 807656004303 807656005218 807656005225
76274-324RL 31.85 USD
76274-324 76274-830 76274-832 76274-834 76274-836 76274-838
Xylene Resistant Resin Ribbons
Printer Ribbons
These resin ribbons for thermal transfer printers will produce a chemical- and xylene-resistant printout, perfect for printing solvent-resistant labels.
  • Thermal transfer xylene and solvent-resistant ribbon
When used with the appropriate label, the ribbon will provide a xylene- and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Used for histology and other applications that involve exposure to harsh chemicals and solvents.