Autoclavable Piggyback Labels
Supplier: GA INTERNATIONAL INC
807656015859 807656015866
76273-688RL 370.63 USD
76273-688 76273-690
Autoclavable Piggyback Labels
Labels
Autoclave-resistant piggyback labels for thermal transfer printers. These removable labels have a unique label-on-label design, perfect for identifying multiple linked items and surfaces.
  • Temperature range: −60 to +150 °C (−76 to +302 °F)
  • Special label-on-label design allows for the identification of multiple subdivided items and surfaces
  • Autoclavable, labels can withstand 20 autoclave cycles
  • Removable adhesive
  • Compatible with all major brands of thermal transfer printers
Removable autoclave-resistant piggyback labels ideal for metal, plastic, and glass surfaces, including surgical instruments, glassware, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Special label on label design allows users to print once and use smaller nested labels later on different surfaces as needed. These durable autoclave thermal transfer labels will not leave any adhesive residue after removal. They are weather- and water-proof, alcohol-resistant, and have been tested to endure 20 autoclave cycles (at 121 °C/250 °F, 16.5 psi for 20 minutes). Compatible with all major brands of thermal-transfer printers.