ABT-491 hydrochloride ≥98% (by HPLC), Sigma-Aldrich®

A9227-5MG A9227-25MG
220104-849EA 394.85 USD
220104-849 220104-848
ABT-491 hydrochloride ≥98% (by HPLC), Sigma-Aldrich®
ABT-491 hydrochloride

Highly potent, selective and orally active platelet-activating factor (PAF) receptor antagonist. Ki = 0.6 nM in human platelets.

Formula: C₂₈H₂₂FN₅O₂·xHCl
Storage Temperature: Refrigerator
MDL Number: MFCD00945301
CAS Number: 189689-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR